missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Piperazinylcarbonyl)-1,4-benzodioxane Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America P189025G

 View more versions of this product

Catalog No. P189025G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-(1-Piperazinylcarbonyl)-1,4-benzodioxane Hydrochloride
263°C
C13H17ClN2O3
25 g
1-[(1,4-Benzodioxan-2-yl)carbonyl]piperazine Hydrochloride
C1CN(CCN1)C(=O)C2COC3=CC=CC=C3O2.Cl
284.74
284.74
Crystalline Powder
70918-74-0
White
MFCD00729051
2811
TUKBWYXLYINULI-UHFFFAOYSA-N
2,3-dihydro-1,4-benzodioxin-3-yl(piperazin-1-yl)methanone;hydrochloride
2822434
≥98.0% (HPLC)

Chemical Identifiers

70918-74-0
284.74
TUKBWYXLYINULI-UHFFFAOYSA-N
2822434
C1CN(CCN1)C(=O)C2COC3=CC=CC=C3O2.Cl
C13H17ClN2O3
MFCD00729051
1-[(1,4-Benzodioxan-2-yl)carbonyl]piperazine Hydrochloride
2,3-dihydro-1,4-benzodioxin-3-yl(piperazin-1-yl)methanone;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions