missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Piperazinylcarbonyl)-1,4-benzodioxane Hydrochloride 98.0+%, TCI America™

Catalog No. p-7135306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18905G 5 g
P189025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18905G Supplier TCI America Supplier No. P18905G

Chemical Identifiers

CAS 70918-74-0
Molecular Formula C13H17ClN2O3
Molecular Weight (g/mol) 284.74
MDL Number MFCD00729051
InChI Key TUKBWYXLYINULI-UHFFFAOYSA-N
Synonym 1-[(1,4-Benzodioxan-2-yl)carbonyl]piperazine Hydrochloride
PubChem CID 2822434
IUPAC Name 2,3-dihydro-1,4-benzodioxin-3-yl(piperazin-1-yl)methanone;hydrochloride
SMILES C1CN(CCN1)C(=O)C2COC3=CC=CC=C3O2.Cl

Specifications

Melting Point 263°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 284.74
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(1-Piperazinylcarbonyl)-1,4-benzodioxane Hydrochloride