missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Piperazinyl)phenol, 98%

Catalog No. AAB2025218
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2025218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2025218 Supplier Thermo Scientific Chemicals Supplier No. B2025218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1011-17-2 | C10H14N2O | 178.235 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1011-17-2
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00190246
InChI Key UORNTHBBLYBAJJ-UHFFFAOYSA-N
Synonym 2-piperazin-1-yl phenol, 1-2-hydroxyphenyl piperazine, 2-1-piperazinyl phenol, 2-1-piperazino phenol, o-1-piperazinyl phenol, 2-piperazin-1-yl-phenol, n-2-hydroxyphenyl piperazine, phenol, 2-1-piperazinyl, 1-2-hydroxyphenyl-piperazine, 2-piperazinylphenol
PubChem CID 70530
IUPAC Name 2-piperazin-1-ylphenol
SMILES C1CN(CCN1)C2=CC=CC=C2O

Specifications

Melting Point 122°C to 127°C
Quantity 50 g
UN Number UN3263
Beilstein 10646
Sensitivity Air Sensitive
Formula Weight 178.24
Percent Purity 98%
Chemical Name or Material 2-(1-Piperazinyl)phenol
Hazard Category H314
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
EINECSNumber 213-782-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only