missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Naphthyl)acetamide 98.0+%, TCI America™

Catalog No. N062425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N062425G 25 g
N0624500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N062425G Supplier TCI America Supplier No. N062425G

Chemical Identifiers

CAS 86-86-2
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.226
MDL Number MFCD00004047
InChI Key XFNJVKMNNVCYEK-UHFFFAOYSA-N
Synonym 1-naphthaleneacetamide, 1-naphthylacetamide, 2-1-naphthyl acetamide, frufix, rootone, naphthaleneacetamide, amid-thin, dirigol n, amid-thin w, naam
PubChem CID 6861
ChEBI CHEBI:81810
IUPAC Name 2-naphthalen-1-ylacetamide
SMILES C1=CC=C2C(=C1)C=CC=C2CC(=O)N

Specifications

Melting Point 184°C
Color White-Yellow
Quantity 25 g
Formula Weight 185.23
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(1-Naphthyl)acetamide
RTECSNumber QJ0590000
TSCA Yes