missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Imidazolyl)acetonitrile, 95%

CAS: 98873-55-3 | C5H5N3 | 107.116 g/mol

$110.86 - $340.70

Chemical Identifiers

CAS 98873-55-3
Molecular Formula C5H5N3
Molecular Weight (g/mol) 107.116
MDL Number MFCD06421433
InChI Key ZPGCVVBPGQJSPX-UHFFFAOYSA-N
Synonym 2-1h-imidazol-1-yl acetonitrile, imidazol-1-yl-acetonitrile, 1-cyanomethyl imidazole, 1h-imidazole-1-acetonitrile, 1h-imidazol-1-ylacetonitrile, 2-1-imidazolyl acetonitrile, 1-imidazolyl acetonitrile, 2-imidazol-1-yl acetonitrile, 1h-imidazol-1-yl acetonitrile, 1-cyanomethylimidazole
PubChem CID 11804695
IUPAC Name 2-imidazol-1-ylacetonitrile
SMILES C1=CN(C=N1)CC#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66012MD
View Documents
Thermo Scientific Chemicals
H66012MD
250 mg
Each for $110.86
Only null left
 
AAH6601203
View Documents
Thermo Scientific Chemicals
H6601203
1 g
Each for $340.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98873-55-3
107.116
ZPGCVVBPGQJSPX-UHFFFAOYSA-N
11804695
C1=CN(C=N1)CC#N
C5H5N3
MFCD06421433
2-1h-imidazol-1-yl acetonitrile, imidazol-1-yl-acetonitrile, 1-cyanomethyl imidazole, 1h-imidazole-1-acetonitrile, 1h-imidazol-1-ylacetonitrile, 2-1-imidazolyl acetonitrile, 1-imidazolyl acetonitrile, 2-imidazol-1-yl acetonitrile, 1h-imidazol-1-yl acetonitrile, 1-cyanomethylimidazole
2-imidazol-1-ylacetonitrile

Specifications

98873-55-3
MFCD06421433
UN3439
2-1h-imidazol-1-yl acetonitrile, imidazol-1-yl-acetonitrile, 1-cyanomethyl imidazole, 1h-imidazole-1-acetonitrile, 1h-imidazol-1-ylacetonitrile, 2-1-imidazolyl acetonitrile, 1-imidazolyl acetonitrile, 2-imidazol-1-yl acetonitrile, 1h-imidazol-1-yl acetonitrile, 1-cyanomethylimidazole
C1=CN(C=N1)CC#N
107.116
107.12
Solid
C5H5N3
250 mg
7473110
ZPGCVVBPGQJSPX-UHFFFAOYSA-N
2-imidazol-1-ylacetonitrile
11804695
95%
2-(1-Imidazolyl)acetonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only