accessibility menu, dialog, popup

UserName

NNN

CAS RN 98873-55-3

IUPAC Name: 2-(1H-imidazol-1-yl)acetonitrile
Synonyms: 2-(imidazol-1-yl)acetonitrile
Molecular Weight (g/mol): 107.12
Molecular Formula: C5H5N3
InChi Key: ZPGCVVBPGQJSPX-UHFFFAOYSA-N
SMILES: N#CCN1C=CN=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)