missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Cyclohexenyl)cyclohexanone 85.0+%, TCI America™

Catalog No. p-7124826
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C116025ML 25 mL
C1160500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C116025ML Supplier TCI America Supplier No. C116025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1502-22-3
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.28
MDL Number MFCD00013796
InChI Key GVNVAWHJIKLAGL-UHFFFAOYNA-N
Synonym 2-1-cyclohexenyl cyclohexanone, chch, 2-cyclohexenylcyclohexanone, 2-1-cyclohexen-1-yl cyclohexan-1-one, 1,1'-bi cyclohexan-1-en-2-one, cyclohexanone, 2-1-cyclohexen-1-yl, 2-cyclohex-1-enyl cyclohexanone, 2-cyclohex-1-en-1-yl cyclohexan-1-one, 2-1-cyclohexen-1-yl cyclohexanone, cyclohexanone, 2-cyclohexenyl
PubChem CID 101175
IUPAC Name 2-(cyclohex-1-en-1-yl)cyclohexan-1-one
SMILES O=C1CCCCC1C1=CCCCC1

Specifications

Color Yellow
Boiling Point 275°C
Quantity 25 mL
Formula Weight 178.28
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(1-Cyclohexenyl)cyclohexanone
EINECSNumber (4)-0027
TSCA Yes