missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Cyclohexenyl)cyclohexanone, 85+%, cont. ca 10% 2-cyclohexylidenecyclohexanone

Catalog No. aal0979814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0979814 25 g
AAL0979822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0979814 Supplier Thermo Scientific Chemicals Supplier No. L0979814
Only null left

CAS: 1502-22-3 | C12H18O | 178.28 g/mol

It is a perfuming agent. It is used for special solvents and as a raw material in agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a perfuming agent. It is used for special solvents and as a raw material in agrochemicals and dyestuffs.

Solubility
Soluble in water. Insoluble in alcohol.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 1502-22-3
Molecular Formula C12H18O
Molecular Weight (g/mol) 178.28
MDL Number MFCD00013796
InChI Key GVNVAWHJIKLAGL-UHFFFAOYNA-N
Synonym 2-1-cyclohexenyl cyclohexanone, chch, 2-cyclohexenylcyclohexanone, 2-1-cyclohexen-1-yl cyclohexan-1-one, 1,1'-bi cyclohexan-1-en-2-one, cyclohexanone, 2-1-cyclohexen-1-yl, 2-cyclohex-1-enyl cyclohexanone, 2-cyclohex-1-en-1-yl cyclohexan-1-one, 2-1-cyclohexen-1-yl cyclohexanone, cyclohexanone, 2-cyclohexenyl
PubChem CID 101175
SMILES O=C1CCCCC1C1=CCCCC1

Specifications

Density 0.998
Boiling Point 265°C
Flash Point >110°C (230°F)
Refractive Index 1.506
Quantity 25 g
Beilstein 1308501
Solubility Information Soluble in water. Insoluble in alcohol.
IUPAC Name 2-(cyclohexen-1-yl)cyclohexan-1-one
Formula Weight 178.28
Percent Purity ≥85%
Assay cont. ≈10% 2-cyclohexylidenecyclohexanone
Chemical Name or Material 2-(1-Cyclohexenyl)cyclohexanone
Show More Show Less
EINECSNumber 216-120-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only