missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Benzofuran-3-yl)ethyl 4-methylbenzenesulfonate, ≥97%, Thermo Scientific™

Catalog No. CC40844DA
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC40844DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC40844DA Supplier Thermo Scientific Chemicals Supplier No. CC40844DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26278-25-1
Molecular Formula C17H16O4S
Molecular Weight (g/mol) 316.37
MDL Number MFCD08271906
InChI Key OEJOFPWUQPRTDR-UHFFFAOYSA-N
Synonym 2-1-benzofuran-3-yl ethyl 4-methylbenzenesulfonate, 3-benzofuranethanol,3-4-methylbenzenesulfonate, p-toluenesulfonic acid 2-benzofuran-3-yl ethyl ester, 2-1-benzofuran-3-yl ethyl 4-methylbenzene-1-sulfonate, 2-1-benzo b furan-3-yl ethyl-4-methylbenzenesulfonate
PubChem CID 19081435
IUPAC Name 2-(1-benzofuran-3-yl)ethyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCC1=COC2=CC=CC=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1g
Formula Weight 316.38
Percent Purity 97%
Chemical Name or Material 2-(1-Benzofuran-3-yl)ethyl 4-methylbenzenesulfonate
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.