accessibility menu, dialog, popup

UserName

CAS RN 26278-25-1

IUPAC Name: 2-(1-benzofuran-3-yl)ethyl 4-methylbenzene-1-sulfonate
Synonyms: 2-(1-benzofuran-3-yl)ethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 316.37
Molecular Formula: C17H16O4S
InChi Key: OEJOFPWUQPRTDR-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCCC1=COC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)