missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Adamantyl)-4-bromoanisole 98.0+%, TCI America™

Catalog No. a232125g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
A232125G 25 g
A23215G 5 g
2 options

Catalog No. A232125G

Supplier: TCI America A232125G

Only null left

Chemical Identifiers

CAS 104224-63-7
Molecular Formula C17H21BrO
Molecular Weight (g/mol) 321.258
MDL Number MFCD03855308
InChI Key QQAMHHZQONQBFZ-UHFFFAOYSA-N
Synonym 1-(5-Bromo-2-methoxyphenyl)adamantane
PubChem CID 3285024
IUPAC Name 1-(5-bromo-2-methoxyphenyl)adamantane
SMILES COC1=C(C=C(C=C1)Br)C23CC4CC(C2)CC(C4)C3

Specifications

Melting Point 144°C
Color White
Quantity 25 g
Formula Weight 321.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(1-Adamantyl)-4-bromoanisole