missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1,3-Dioxolan-2-yl)ethyltriphenylphosphonium Bromide 97.0+%, TCI America™
Supplier: TCI America D205625G
Specifications
| 2-(1,3-Dioxolan-2-yl)ethyltriphenylphosphonium Bromide | |
| 155°C | |
| MFCD00075119 | |
| 2-1,3-dioxolan-2-yl ethyltriphenylphosphonium bromide, 2-1,3-dioxolan-2-yl ethyl triphenylphosphonium bromide, phosphonium, 2-1,3-dioxolan-2-yl ethyl triphenyl-, bromide, 2-1,3-dioxolan-2-yl ethyl triphenylphosphanium bromide, acmc-1bl1m, 2-1,3-dioxolan-2-yl ethyltrip, phosphonium, 2-1,3-dioxolan-2-yl ethyl triphenyl-, bromide 1:1, 2-1,3-dioxolan-2-yl ethyltriphenylphosp, 2-1,3-dioxolan-2-yl ethyltriphenylphosphoniumbromide | |
| C1COC(O1)CC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-] | |
| 443.321 | |
| 443.32 | |
| Crystalline Powder |
| 86608-70-0 | |
| C23H24BrO2P | |
| 25 g | |
| ZCJKBPSRKLHANV-UHFFFAOYSA-M | |
| 2-(1,3-dioxolan-2-yl)ethyl-triphenylphosphanium;bromide | |
| 2733834 | |
| ≥97.0% (T) |
Chemical Identifiers
| 86608-70-0 | |
| 443.321 | |
| ZCJKBPSRKLHANV-UHFFFAOYSA-M | |
| 2733834 | |
| C1COC(O1)CC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Br-] |
| C23H24BrO2P | |
| MFCD00075119 | |
| 2-1,3-dioxolan-2-yl ethyltriphenylphosphonium bromide, 2-1,3-dioxolan-2-yl ethyl triphenylphosphonium bromide, phosphonium, 2-1,3-dioxolan-2-yl ethyl triphenyl-, bromide, 2-1,3-dioxolan-2-yl ethyl triphenylphosphanium bromide, acmc-1bl1m, 2-1,3-dioxolan-2-yl ethyltrip, phosphonium, 2-1,3-dioxolan-2-yl ethyl triphenyl-, bromide 1:1, 2-1,3-dioxolan-2-yl ethyltriphenylphosp, 2-1,3-dioxolan-2-yl ethyltriphenylphosphoniumbromide | |
| 2-(1,3-dioxolan-2-yl)ethyl-triphenylphosphanium;bromide |
Safety and Handling
TSCA : Yes