missing translation for 'onlineSavingsMsg'
Learn More

2-(1,3-Dioxan-2-yl)ethyltriphenylphosphonium Bromide 98.0+%, TCI America™

Catalog No. p-7126855
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16555G 5 g
D165525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D16555G Supplier TCI America Supplier No. D16555G
Only null left

Chemical Identifiers

CAS 69891-92-5
Molecular Formula C24H26BrO2P
Molecular Weight (g/mol) 457.35
MDL Number MFCD00012000
InChI Key XETDBHNHTOJWPZ-UHFFFAOYSA-M
Synonym 2-1,3-dioxan-2-yl ethyltriphenylphosphonium bromide, 2-1,3-dioxan-2-yl ethyl triphenylphosphonium bromide, 2-1,3-dioxan-2-yl ethyl triphenylphosphanium bromide, acmc-209obj, xqaupnaycikwct-uhfffaoysa-m, 1,3-dioxan-2-ylethyltriphenylphosphinebromide, 2-1,3-dioxan-2-yl ethyltriphenylphosphonium bromi, 2-1,3-dioxan-2-yl ethyltriphenylphosphoniumbromide, 2-1,3-dioxane-2-yl ethyltriphenylphosphonium bromide
PubChem CID 2724168
IUPAC Name [2-(1,3-dioxan-2-yl)ethyl]triphenylphosphanium bromide
SMILES [Br-].C(C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1OCCCO1

Specifications

Melting Point 189°C
Color White-Yellow
Quantity 5 g
Formula Weight 457.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(1,3-Dioxan-2-yl)ethyltriphenylphosphonium Bromide
EINECSNumber (5)-6416
TSCA No