missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1,3-Dioxan-2-yl)ethyltriphenylphosphonium Bromide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 189°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 457.35 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-(1,3-Dioxan-2-yl)ethyltriphenylphosphonium Bromide |
Safety and Handling
| EINECSNumber | (5)-6416 |
| TSCA | No |