missing translation for 'onlineSavingsMsg'
Learn More

[2-(1,3-Dioxan-2-yl)ethyl]triphenylphosphonium bromide, 98+%

Catalog No. p-7044156
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0490906 5 g
AAL0490914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0490906 Supplier Thermo Scientific Chemicals Supplier No. L0490906
Only null left

CAS: 69891-92-5 | C24H26BrO2P | 457.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 69891-92-5
Molecular Formula C24H26BrO2P
Molecular Weight (g/mol) 457.35
MDL Number MFCD00012000
InChI Key XETDBHNHTOJWPZ-UHFFFAOYSA-M
Synonym 2-1,3-dioxan-2-yl ethyltriphenylphosphonium bromide, 2-1,3-dioxan-2-yl ethyl triphenylphosphonium bromide, 2-1,3-dioxan-2-yl ethyl triphenylphosphanium bromide, acmc-209obj, xqaupnaycikwct-uhfffaoysa-m, 1,3-dioxan-2-ylethyltriphenylphosphinebromide, 2-1,3-dioxan-2-yl ethyltriphenylphosphonium bromi, 2-1,3-dioxan-2-yl ethyltriphenylphosphoniumbromide, 2-1,3-dioxane-2-yl ethyltriphenylphosphonium bromide
PubChem CID 2724168
IUPAC Name 2-(1,3-dioxan-2-yl)ethyl-triphenylphosphanium;bromide
SMILES [Br-].C(C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1OCCCO1

Specifications

Melting Point 183°C to 195°C
Quantity 5 g
Beilstein 4117571
Sensitivity Moisture sensitive
Formula Weight 457.36
Percent Purity ≥98%
Chemical Name or Material [2-(1,3-Dioxan-2-yl)ethyl]triphenylphosphonium bromide
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 274-189-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only