missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,1,3-Benzoxadiazole-5-carboxylic acid, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 19155-88-5 |
|---|---|
| Molecular Formula | C7H3N2O3 |
| Molecular Weight (g/mol) | 163.11 |
| MDL Number | MFCD00276977 |
| InChI Key | WZUFYJFTOVGJJT-UHFFFAOYSA-M |
| Synonym | benzo c 1,2,5 oxadiazole-5-carboxylic acid, benzofurazan-5-carboxylic acid, 5-benzofurazancarboxylic acid, benzo c 1,2,5-oxadiazole-5-carboxylic acid, pubchem3995, maybridge1_002587, 4-bromo-2,6-dimethylanisole,, 2,1,3-benzoxadiazole-5-carboxylicacid, benzo 1,2,5 oxadiazole-5-carboxylic acid, benzo 2,1,3 oxadiazole-5-carboxylic acid |
| PubChem CID | 2735454 |
| IUPAC Name | 2,1,3-benzoxadiazole-5-carboxylic acid |
| SMILES | [O-]C(=O)C1=CC2=NON=C2C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
CC09701CB
|
Thermo Scientific Chemicals
CC09701CB |
250mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC09701DA
|
Thermo Scientific Chemicals
CC09701DA |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC09701EA
|
Thermo Scientific Chemicals
CC09701EA |
10g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 19155-88-5 | |
| 163.11 | |
| WZUFYJFTOVGJJT-UHFFFAOYSA-M | |
| 2735454 | |
| [O-]C(=O)C1=CC2=NON=C2C=C1 |
| C7H3N2O3 | |
| MFCD00276977 | |
| benzo c 1,2,5 oxadiazole-5-carboxylic acid, benzofurazan-5-carboxylic acid, 5-benzofurazancarboxylic acid, benzo c 1,2,5-oxadiazole-5-carboxylic acid, pubchem3995, maybridge1_002587, 4-bromo-2,6-dimethylanisole,, 2,1,3-benzoxadiazole-5-carboxylicacid, benzo 1,2,5 oxadiazole-5-carboxylic acid, benzo 2,1,3 oxadiazole-5-carboxylic acid | |
| 2,1,3-benzoxadiazole-5-carboxylic acid |
Specifications
| 19155-88-5 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD00276977 | |
| benzo c 1,2,5 oxadiazole-5-carboxylic acid, benzofurazan-5-carboxylic acid, 5-benzofurazancarboxylic acid, benzo c 1,2,5-oxadiazole-5-carboxylic acid, pubchem3995, maybridge1_002587, 4-bromo-2,6-dimethylanisole,, 2,1,3-benzoxadiazole-5-carboxylicacid, benzo 1,2,5 oxadiazole-5-carboxylic acid, benzo 2,1,3 oxadiazole-5-carboxylic acid | |
| [O-]C(=O)C1=CC2=NON=C2C=C1 | |
| 163.11 | |
| 164.12 | |
| 2, 1, 3-Benzoxadiazole-5-carboxylic acid |
| 96.5 | |
| 97% | |
| C7H3N2O3 | |
| 250mg | |
| WZUFYJFTOVGJJT-UHFFFAOYSA-M | |
| 2,1,3-benzoxadiazole-5-carboxylic acid | |
| 2735454 | |
| 97% |