missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,1,3-Benzoxadiazole-5-carboxylic acid, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC09701CB
Specifications
| 2, 1, 3-Benzoxadiazole-5-carboxylic acid | |
| 96.5 | |
| 97% | |
| C7H3N2O3 | |
| 250mg | |
| WZUFYJFTOVGJJT-UHFFFAOYSA-M | |
| 2,1,3-benzoxadiazole-5-carboxylic acid | |
| 2735454 | |
| 97% |
| 19155-88-5 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD00276977 | |
| benzo c 1,2,5 oxadiazole-5-carboxylic acid, benzofurazan-5-carboxylic acid, 5-benzofurazancarboxylic acid, benzo c 1,2,5-oxadiazole-5-carboxylic acid, pubchem3995, maybridge1_002587, 4-bromo-2,6-dimethylanisole,, 2,1,3-benzoxadiazole-5-carboxylicacid, benzo 1,2,5 oxadiazole-5-carboxylic acid, benzo 2,1,3 oxadiazole-5-carboxylic acid | |
| [O-]C(=O)C1=CC2=NON=C2C=C1 | |
| 163.11 | |
| 164.12 |
Chemical Identifiers
| 19155-88-5 | |
| 163.11 | |
| WZUFYJFTOVGJJT-UHFFFAOYSA-M | |
| 2735454 | |
| [O-]C(=O)C1=CC2=NON=C2C=C1 |
| C7H3N2O3 | |
| MFCD00276977 | |
| benzo c 1,2,5 oxadiazole-5-carboxylic acid, benzofurazan-5-carboxylic acid, 5-benzofurazancarboxylic acid, benzo c 1,2,5-oxadiazole-5-carboxylic acid, pubchem3995, maybridge1_002587, 4-bromo-2,6-dimethylanisole,, 2,1,3-benzoxadiazole-5-carboxylicacid, benzo 1,2,5 oxadiazole-5-carboxylic acid, benzo 2,1,3 oxadiazole-5-carboxylic acid | |
| 2,1,3-benzoxadiazole-5-carboxylic acid |