missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzoxadiazol-4-ylmethanol, Thermo Scientific™

Catalog No. p-3075350
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC09609DA 1g
CC09609DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC09609DA Supplier Thermo Scientific Chemicals Supplier No. CC09609DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 175609-19-5
Molecular Formula C7H6N2O2
Molecular Weight (g/mol) 150.14
MDL Number MFCD04115376
InChI Key OKDMFOBEJCALFD-UHFFFAOYSA-N
Synonym 4-hydroxymethylbenzofurazan, benzo c 1,2,5 oxadiazol-4-ylmethanol, 4-hydroxymethyl-benzo c 1,2,5 oxadiazole, 2,1,3-benzoxadiazole-4-methanol, 1,2,3-benzoxadiazol-4-ylmethanol, benzo c 1,2,5-oxadiazol-4-ylmethan-1-ol
PubChem CID 2795215
IUPAC Name 2,1,3-benzoxadiazol-4-ylmethanol
SMILES OCC1=CC=CC2=NON=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1g
Formula Weight 150.14
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzoxadiazol-4-ylmethanol