missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazole 99.0+%, TCI America™

Catalog No. p-7118744
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B092110G 10 g
B092125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B092110G Supplier TCI America Supplier No. B092110G
Only null left

Chemical Identifiers

CAS 273-13-2
Molecular Formula C6H4N2S
Molecular Weight (g/mol) 136.172
MDL Number MFCD00005809
InChI Key PDQRQJVPEFGVRK-UHFFFAOYSA-N
Synonym piazthiole, benzisothiadiazole, benzo 1,2,5 thiadiazole, benzo c 1,2,5 thiadiazole, 2-thia-1,3-diaza-2h-isoindene, 3,4-benzo-1,2,5-thiadiazole, 2??,1,3-benzothiadiazole, 2,3-benzothiadiazole, 3,2,5-thiadiazole, benzo-1,2,5-thiadiazole
PubChem CID 67502
IUPAC Name 2,1,3-benzothiadiazole
SMILES C1=CC2=NSN=C2C=C1

Specifications

Melting Point 44°C
Color White-Yellow
Boiling Point 64°C
Quantity 10 g
Formula Weight 136.17
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,1,3-Benzothiadiazole