accessibility menu, dialog, popup

UserName

CAS RN 273-13-2

IUPAC Name: 2,1,3-benzothiadiazole
Synonyms: 2,1,3-benzothiadiazole
Molecular Weight (g/mol): 136.17
Molecular Formula: C6H4N2S
InChi Key: PDQRQJVPEFGVRK-UHFFFAOYSA-N
SMILES: S1N=C2C=CC=CC2=N1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)