missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazole, 98%, Thermo Scientific Chemicals

$128.86 - $128.86

Chemical Identifiers

CAS 273-13-2
Molecular Formula C6H4N2S
Molecular Weight (g/mol) 136.172
MDL Number MFCD00005809
InChI Key PDQRQJVPEFGVRK-UHFFFAOYSA-N
Synonym piazthiole, benzisothiadiazole, benzo 1,2,5 thiadiazole, benzo c 1,2,5 thiadiazole, 2-thia-1,3-diaza-2h-isoindene, 3,4-benzo-1,2,5-thiadiazole, 2??,1,3-benzothiadiazole, 2,3-benzothiadiazole, 3,2,5-thiadiazole, benzo-1,2,5-thiadiazole
PubChem CID 67502
IUPAC Name 2,1,3-benzothiadiazole
SMILES C1=CC2=NSN=C2C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4373009
View Documents
Thermo Scientific Chemicals
04373009
10 g N/A
Only null left
 
Description

Description

The product is used as an industrial solvent, and pharmaceutical intermediates for organic synthesis and used nitrocellulose and other resin, wax, fatty, dyes and other solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

273-13-2
136.172
PDQRQJVPEFGVRK-UHFFFAOYSA-N
67502
C1=CC2=NSN=C2C=C1
C6H4N2S
MFCD00005809
piazthiole, benzisothiadiazole, benzo 1,2,5 thiadiazole, benzo c 1,2,5 thiadiazole, 2-thia-1,3-diaza-2h-isoindene, 3,4-benzo-1,2,5-thiadiazole, 2??,1,3-benzothiadiazole, 2,3-benzothiadiazole, 3,2,5-thiadiazole, benzo-1,2,5-thiadiazole
2,1,3-benzothiadiazole

Specifications

273-13-2
206°C
C6H4N2S
10 g
PDQRQJVPEFGVRK-UHFFFAOYSA-N
2,1,3-benzothiadiazole
67502
98%
2,1,3-Benzothiadiazole
42°C to 44°C
95°C (203°F)
MFCD00005809
piazthiole, benzisothiadiazole, benzo 1,2,5 thiadiazole, benzo c 1,2,5 thiadiazole, 2-thia-1,3-diaza-2h-isoindene, 3,4-benzo-1,2,5-thiadiazole, 2??,1,3-benzothiadiazole, 2,3-benzothiadiazole, 3,2,5-thiadiazole, benzo-1,2,5-thiadiazole
C1=CC2=NSN=C2C=C1
136.172
136.18
Powder
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 205-985-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only