Learn More
(1S)-(-)-alpha-Pinene, 98%
CAS: 7785-26-4 | C10H16 | 136.24 g/mol
Supplier: Thermo Scientific Chemicals 131270025
| Quantity | 2.5 L |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7785-26-4 | |
| 136.24 | |
| --alpha-pinene, 1s,5s-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1s---alpha-pinene, unii-tzr3gm95pr, dl-alpha-pinene, alpha-pinene,-, 1s,5s-alpha-pinene, tzr3gm95pr, 1s,5s-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1s,5s | |
| CHEBI:28660 | |
| CC1=CCC2CC1C2(C)C |
| C10H16 | |
| GRWFGVWFFZKLTI-IUCAKERBSA-N | |
| 440968 | |
| (1S,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-ene |
Specifications
| (1S)-(-)-α-Pinene | |
| -64.0°C | |
| 0.8595g/mL | |
| 33°C | |
| 97.5% min. (GC) | |
| C10H16 | |
| 2.5 L | |
| 0.8595 | |
| - 41.50 (20.00°C c=neat) | |
| --alpha-pinene, 1s,5s-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1s---alpha-pinene, unii-tzr3gm95pr, dl-alpha-pinene, alpha-pinene,-, 1s,5s-alpha-pinene, tzr3gm95pr, 1s,5s-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1s,5s | |
| GRWFGVWFFZKLTI-IUCAKERBSA-N | |
| (1S,5S)-4,6,6-trimethylbicyclo[3.1.1]hept-3-ene | |
| 440968 | |
| 136.24 | |
| Liquid |
| 7785-26-4 | |
| Colorless | |
| 155.0°C to 156.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4650 to 1.4670 | |
| 05,146 | |
| 14,7445 | |
| − 41.50 | |
| Solubility in water: insoluble. Other solubilities: soluble in chloroform,alcohol and ether | |
| CC1=CCC2CC1C2(C)C | |
| 136.24 | |
| CHEBI:28660 | |
| 98% |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Harmful if swallowed.
May cause an allergic skin reaction.
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting
GHS P Statement
Wear protective gloves/protective clothing.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
If skin irritation or rash occurs: Get medical advice/attention.
IF ON SKIN (or hair): Take off immediately all contam
GHS Signal Word: Danger
EINECSNumber : 232-077-3