missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(+)-Menthyl acetate, 99%

CAS: 5157-89-1 | C12H22O2 | 198.306 g/mol

$61.78 - $156.75

Chemical Identifiers

CAS 5157-89-1
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00062977
InChI Key XHXUANMFYXWVNG-DLOVCJGASA-N
Synonym neoisomenthyl acetate, +-menthylacetate, 1s-2alpha-1-methylethyl-5alpha-methylcyclohexan-1alpha-ol acetate
PubChem CID 21630934
IUPAC Name [(1S,2S,5S)-5-methyl-2-propan-2-ylcyclohexyl] acetate
SMILES CC1CCC(C(C1)OC(=O)C)C(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL13987MD
View Documents
Thermo Scientific Chemicals
L13987MD
250 mg
Each for $61.78
Only null left
 
AAL1398703
View Documents
Thermo Scientific Chemicals
L1398703
1 g
Each for $156.75
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5157-89-1
198.306
XHXUANMFYXWVNG-DLOVCJGASA-N
21630934
CC1CCC(C(C1)OC(=O)C)C(C)C
C12H22O2
MFCD00062977
neoisomenthyl acetate, +-menthylacetate, 1s-2alpha-1-methylethyl-5alpha-methylcyclohexan-1alpha-ol acetate
[(1S,2S,5S)-5-methyl-2-propan-2-ylcyclohexyl] acetate

Specifications

5157-89-1
229°C to 230°C
C12H22O2
MFCD00062977
1909419
XHXUANMFYXWVNG-DLOVCJGASA-N
CC1CCC(C(C1)OC(=O)C)C(C)C
198.306
198.31
(1S)-(+)-Menthyl acetate
0.925
77°C (170°F)
1.447
250 mg
neoisomenthyl acetate, +-menthylacetate, 1s-2alpha-1-methylethyl-5alpha-methylcyclohexan-1alpha-ol acetate
+80.5° (c=8 in Benzene)
[(1S,2S,5S)-5-methyl-2-propan-2-ylcyclohexyl] acetate
21630934
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only