missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S)-(+)-Camphorquinone 97.0+%, TCI America™
$106.25 - $322.48
Chemical Identifiers
| CAS | 2767-84-2 |
|---|---|
| Molecular Formula | C10H14O2 |
| Molecular Weight (g/mol) | 166.22 |
| MDL Number | MFCD00064159 |
| InChI Key | VNQXSTWCDUXYEZ-LHLIQPBNSA-N |
| Synonym | (1S)-(+)-2,3-Bornanedione |
| PubChem CID | 12222796 |
| IUPAC Name | (1S,4S)-4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione |
| SMILES | CC1(C2CCC1(C(=O)C2=O)C)C |
Chemical Identifiers
| 2767-84-2 | |
| 166.22 | |
| VNQXSTWCDUXYEZ-LHLIQPBNSA-N | |
| 12222796 | |
| CC1(C2CCC1(C(=O)C2=O)C)C |
| C10H14O2 | |
| MFCD00064159 | |
| (1S)-(+)-2,3-Bornanedione | |
| (1S,4S)-4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione |
Specifications
| 2767-84-2 | |
| White-Yellow | |
| MFCD00064159 | |
| (1S)-(+)-2,3-Bornanedione | |
| CC1(C2CCC1(C(=O)C2=O)C)C | |
| 166.22 | |
| 166.22 | |
| Crystalline Powder |
| 200°C | |
| C10H14O2 | |
| 1 g | |
| VNQXSTWCDUXYEZ-LHLIQPBNSA-N | |
| (1S,4S)-4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione | |
| 12222796 | |
| ≥97.0% (GC) | |
| (1S)-(+)-Camphorquinone |
Safety and Handling
TSCA : Yes