accessibility menu, dialog, popup

UserName

CAS RN 2767-84-2

H3CH3CCH3OO

CAS RN 2767-84-2

IUPAC Name: 1,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
Synonyms: camphorquinone camphoroquinone campherquinone camphoquinone bornanedione camphor quinone
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: VNQXSTWCDUXYEZ-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC1(C)C(=O)C2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results