missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(-)-beta-Pinene, 99%

CAS: 18172-67-3 | C10H16 | 136.238 g/mol

$57.97 - $431.75

Chemical Identifiers

CAS 18172-67-3
Molecular Formula C10H16
Molecular Weight (g/mol) 136.238
MDL Number MFCD00001345
InChI Key WTARULDDTDQWMU-IUCAKERBSA-N
Synonym --beta-pinene, --nopinene, 1s,5s-6,6-dimethyl-2-methylenebicyclo 3.1.1 heptane, 1s---beta-pinene, --b-pinene, --pin-2 10-ene, unii-afn153a7su, --?-pinene, 1s-1-beta-pinene, afn153a7su
PubChem CID 440967
ChEBI CHEBI:28359
IUPAC Name (1S,5S)-6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane
SMILES CC1(C2CCC(=C)C1C2)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA17818AE
View Documents Promotion Details
N/A 100 mL
Each for $57.97
Only null left
 
AAA17818AP
View Documents Promotion Details
N/A 500 mL
Each for $138.25
Only null left
 
AAA178180F
View Documents Promotion Details
N/A 2500 mL
Each for $431.75
Only null left
 
Description

Description

It is a useful biochemical for proteomics research. It is also employed in the preparation of natural products, such as grandisol and robustadial. It is also applied as perfuming agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful biochemical for proteomics research. It is also employed in the preparation of natural products, such as grandisol and robustadial. It is also applied as perfuming agent in cosmetics industry.

Solubility
Not miscible or difficult to mix in water, 7.061 mg/L @ 25°C (est). Soluble in alcohol, chloroform and ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources.
Specifications

Chemical Identifiers

18172-67-3
136.238
WTARULDDTDQWMU-IUCAKERBSA-N
440967
(1S,5S)-6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane
C10H16
MFCD00001345
--beta-pinene, --nopinene, 1s,5s-6,6-dimethyl-2-methylenebicyclo 3.1.1 heptane, 1s---beta-pinene, --b-pinene, --pin-2 10-ene, unii-afn153a7su, --?-pinene, 1s-1-beta-pinene, afn153a7su
CHEBI:28359
CC1(C2CCC(=C)C1C2)C

Specifications

18172-67-3
0.86
36°C (96°F)
C10H16
MFCD00001345
2038282
--beta-pinene, --nopinene, 1s,5s-6,6-dimethyl-2-methylenebicyclo 3.1.1 heptane, 1s---beta-pinene, --b-pinene, --pin-2 10-ene, unii-afn153a7su, --?-pinene, 1s-1-beta-pinene, afn153a7su
WTARULDDTDQWMU-IUCAKERBSA-N
CC1(C2CCC(=C)C1C2)C
136.238
CHEBI:28359
99%
-61°C
164°C to 165°C
Turpentine-like
1.478
UN2319
14,7446
Not miscible or difficult to mix in water,7.061mg/L @ 25°C (est). Soluble in alcohol,chloroform and ether.
−21° (Neat)
(1S,5S)-6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane
440967
136.24
(1S)-(-)-beta-Pinene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P261-P264b-P272-P280-P301+P310-P303+P361+P353-P305+P351+P338-P331-P333+P313-P363-P370+P378q-P501c

H226-H304-H315-H317-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TERPENE HYDROCARBONS, N.O.S.

EINECSNumber : 242-060-2

RTECSNumber : DT5077000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only