missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(-)-beta-Pinene, 99%

Catalog No. AAA17818AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA17818AE 100 mL
AAA17818AP 500 mL
AAA178180F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA17818AE Supplier Thermo Scientific Chemicals Supplier No. A17818AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 18172-67-3 | C10H16 | 136.238 g/mol

It is a useful biochemical for proteomics research. It is also employed in the preparation of natural products, such as grandisol and robustadial. It is also applied as perfuming agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful biochemical for proteomics research. It is also employed in the preparation of natural products, such as grandisol and robustadial. It is also applied as perfuming agent in cosmetics industry.

Solubility
Not miscible or difficult to mix in water, 7.061 mg/L @ 25°C (est). Soluble in alcohol, chloroform and ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources.

Chemical Identifiers

CAS 18172-67-3
Molecular Formula C10H16
Molecular Weight (g/mol) 136.238
MDL Number MFCD00001345
InChI Key WTARULDDTDQWMU-IUCAKERBSA-N
Synonym --beta-pinene, --nopinene, 1s,5s-6,6-dimethyl-2-methylenebicyclo 3.1.1 heptane, 1s---beta-pinene, --b-pinene, --pin-2 10-ene, unii-afn153a7su, --?-pinene, 1s-1-beta-pinene, afn153a7su
PubChem CID 440967
ChEBI CHEBI:28359
IUPAC Name (1S,5S)-6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane
SMILES CC1(C2CCC(=C)C1C2)C

Specifications

Melting Point -61°C
Density 0.86
Boiling Point 164°C to 165°C
Flash Point 36°C (96°F)
Odor Turpentine-like
Refractive Index 1.478
Quantity 100 mL
UN Number UN2319
Beilstein 2038282
Merck Index 14,7446
Solubility Information Not miscible or difficult to mix in water,7.061mg/L @ 25°C (est). Soluble in alcohol,chloroform and ether.
Optical Rotation −21° (Neat)
Formula Weight 136.24
Percent Purity 99%
Chemical Name or Material (1S)-(-)-beta-Pinene
Show More Show Less
Hazard Category H226-H304-H315-H317-H319
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P272-P280-P301+P310-P303+P361+P353-P305+P351+P338-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TERPENE HYDROCARBONS, N.O.S.
EINECSNumber 242-060-2
RTECSNumber DT5077000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only