missing translation for 'onlineSavingsMsg'
Learn More

(1S)-(-)-beta-Pinene, 99%

CAS: 18172-67-3 | C10H16 | 136.238 g/mol

Supplier:  Thermo Scientific Chemicals A178180F

 View more versions of this product

Catalog No. AAA178180F


Only null left
Explore available promotions

Description

Description

It is a useful biochemical for proteomics research. It is also employed in the preparation of natural products, such as grandisol and robustadial. It is also applied as perfuming agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a useful biochemical for proteomics research. It is also employed in the preparation of natural products, such as grandisol and robustadial. It is also applied as perfuming agent in cosmetics industry.

Solubility
Not miscible or difficult to mix in water, 7.061 mg/L @ 25°C (est). Soluble in alcohol, chloroform and ether.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources.
Specifications

Specifications

(1S)-(-)-beta-Pinene
-61°C
164°C to 165°C
Turpentine-like
1.478
2500 mL
2038282
--beta-pinene, --nopinene, 1s,5s-6,6-dimethyl-2-methylenebicyclo 3.1.1 heptane, 1s---beta-pinene, --b-pinene, --pin-2 10-ene, unii-afn153a7su, --?-pinene, 1s-1-beta-pinene, afn153a7su
WTARULDDTDQWMU-IUCAKERBSA-N
CC1(C2CCC(=C)C1C2)C
136.238
CHEBI:28359
99%
18172-67-3
0.86
36°C (96°F)
C10H16
MFCD00001345
UN2319
14,7446
Not miscible or difficult to mix in water,7.061mg/L @ 25°C (est). Soluble in alcohol,chloroform and ether.
−21° (Neat)
(1S,5S)-6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane
440967
136.24

Chemical Identifiers

18172-67-3
136.238
WTARULDDTDQWMU-IUCAKERBSA-N
440967
(1S,5S)-6,6-dimethyl-4-methylidenebicyclo[3.1.1]heptane
C10H16
MFCD00001345
--beta-pinene, --nopinene, 1s,5s-6,6-dimethyl-2-methylenebicyclo 3.1.1 heptane, 1s---beta-pinene, --b-pinene, --pin-2 10-ene, unii-afn153a7su, --?-pinene, 1s-1-beta-pinene, afn153a7su
CHEBI:28359
CC1(C2CCC(=C)C1C2)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P240-P241-P242-P243-P261-P264b-P272-P280-P301+P310-P303+P361+P353-P305+P351+P338-P331-P333+P313-P363-P370+P378q-P501c

H226-H304-H315-H317-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TERPENE HYDROCARBONS, N.O.S.

EINECSNumber : 242-060-2

RTECSNumber : DT5077000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only