missing translation for 'onlineSavingsMsg'
Learn More

(1S,6S)-2,8-Diazabicyclo[4.3.0]nonane 98.0+%, TCI America™

Catalog No. D42105G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42105G 5 g
D42101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42105G Supplier TCI America Supplier No. D42105G
Only null left

Chemical Identifiers

CAS 151213-40-0
Molecular Formula C7H14N2
Molecular Weight (g/mol) 126.203
MDL Number MFCD08458306
InChI Key KSCPLKVBWDOSAI-NKWVEPMBSA-N
Synonym (4aS,7aS)-Octahydro-1H-pyrrolo[3,4-b]pyridine
PubChem CID 10313138
IUPAC Name (4aS,7aS)-2,3,4,4a,5,6,7,7a-octahydro-1H-pyrrolo[3,4-b]pyridine
SMILES C1CC2CNCC2NC1

Specifications

Color Yellow
Boiling Point 59°C
Quantity 5 g
Formula Weight 126.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (1S,6S)-2,8-Diazabicyclo[4.3.0]nonane
TSCA No
Recommended Storage Freezer