CAS RN 151213-40-0
IUPAC Name:
octahydro-1H-pyrrolo[3,4-b]pyridine
Synonyms:
octahydro-1H-pyrrolo[3,4-b]pyridine
Molecular Weight (g/mol):
126.2
Molecular Formula:
C7H14N2
InChi Key:
KSCPLKVBWDOSAI-UHFFFAOYNA-N
SMILES:
C1NCC2NCCCC12
1
–
1
of
1
results
1
(1S,6S)-2,8-Diazabicyclo[4.3.0]nonane 98.0+%, TCI America™
CAS: 151213-40-0 Molecular Formula: C7H14N2 Molecular Weight (g/mol): 126.203 MDL Number: MFCD08458306 InChI Key: KSCPLKVBWDOSAI-NKWVEPMBSA-N Synonym: (4aS,7aS)-Octahydro-1H-pyrrolo[3,4-b]pyridine PubChem CID: 10313138 IUPAC Name: (4aS,7aS)-2,3,4,4a,5,6,7,7a-octahydro-1H-pyrrolo[3,4-b]pyridine SMILES: C1CC2CNCC2NC1