missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,6S)-2,8-Diazabicyclo[4.3.0]nonane 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 59°C |
| Quantity | 1 g |
| Formula Weight | 126.20 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | (1S,6S)-2,8-Diazabicyclo[4.3.0]nonane |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |