missing translation for 'onlineSavingsMsg'
Learn More

(1S,4S)-2-Benzyl-2,5-diazabicyclo[2.2.1]heptane Dihydrobromide 98.0+%, TCI America™

Catalog No. B20545G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20545G 5 g
B20541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20545G Supplier TCI America Supplier No. B20545G

Chemical Identifiers

CAS 116258-17-4
Molecular Formula C12H18Br2N2
Molecular Weight (g/mol) 350.098
MDL Number MFCD01321292
InChI Key SOMPEQIPSQFVMO-AQEKLAMFSA-N
PubChem CID 14612542
IUPAC Name (1S,4S)-2-benzyl-2,5-diazabicyclo[2.2.1]heptane;dihydrobromide
SMILES C1C2CNC1CN2CC3=CC=CC=C3.Br.Br

Specifications

Color White
Quantity 5 g
Formula Weight 350.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (1S,4S)-2-Benzyl-2,5-diazabicyclo[2.2.1]heptane Dihydrobromide