Learn More
(1S,3R)-(+)-3-Aminocyclopentanecarboxylic acid, 95%
CAS: 71830-07-4 | C6H11NO2 | 129.159 g/mol
Supplier: Thermo Scientific Chemicals H5204003
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (1S,3R)-(+)-3-Aminocyclopentanecarboxylic acid | |
| C6H11NO2 | |
| 1s,3r-3-aminocyclopentanecarboxylic acid, 1s,3r-3-aminocyclopentane-1-carboxylic acid, +-1s,3r-3-aminocyclopentanecarboxylic acid, 1s,3r-3-amino-cyclopentanecarboxylic acid, cis-3-aminocyclopentanecarboxylic acid, 1s,3r-+-3-aminocyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-amino-, 1s,3r, pubchem18730, +-cacp, cis-3-aminocyclopentane-1-carboxylic acid | |
| C1CC(CC1C(=O)O)N | |
| 1 g | |
| 1502034 | |
| Air Sensitive |
| 71830-07-4 | |
| MFCD00211290 | |
| MLLSSTJTARJLHK-CRCLSJGQSA-N | |
| (1S,3R)-3-aminocyclopentane-1-carboxylic acid | |
| 129.159 | |
| 129.16 | |
| 95% |
Chemical Identifiers
| 71830-07-4 | |
| 129.159 | |
| MLLSSTJTARJLHK-CRCLSJGQSA-N | |
| 1502034 | |
| C1CC(CC1C(=O)O)N |
| C6H11NO2 | |
| MFCD00211290 | |
| 1s,3r-3-aminocyclopentanecarboxylic acid, 1s,3r-3-aminocyclopentane-1-carboxylic acid, +-1s,3r-3-aminocyclopentanecarboxylic acid, 1s,3r-3-amino-cyclopentanecarboxylic acid, cis-3-aminocyclopentanecarboxylic acid, 1s,3r-+-3-aminocyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-amino-, 1s,3r, pubchem18730, +-cacp, cis-3-aminocyclopentane-1-carboxylic acid | |
| (1S,3R)-3-aminocyclopentane-1-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only