missing translation for 'onlineSavingsMsg'
Learn More

(1S,3R)-(+)-3-Aminocyclopentanecarboxylic acid, 95%

Catalog No. p-7034358
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52040MD 250 mg
AAH5204003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52040MD Supplier Thermo Scientific Chemicals Supplier No. H52040MD

CAS: 71830-07-4 | C6H11NO2 | 129.159 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71830-07-4
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00211290
InChI Key MLLSSTJTARJLHK-CRCLSJGQSA-N
Synonym 1s,3r-3-aminocyclopentanecarboxylic acid, 1s,3r-3-aminocyclopentane-1-carboxylic acid, +-1s,3r-3-aminocyclopentanecarboxylic acid, 1s,3r-3-amino-cyclopentanecarboxylic acid, cis-3-aminocyclopentanecarboxylic acid, 1s,3r-+-3-aminocyclopentanecarboxylic acid, cyclopentanecarboxylic acid, 3-amino-, 1s,3r, pubchem18730, +-cacp, cis-3-aminocyclopentane-1-carboxylic acid
PubChem CID 1502034
IUPAC Name (1S,3R)-3-aminocyclopentane-1-carboxylic acid
SMILES C1CC(CC1C(=O)O)N

Specifications

Quantity 250 mg
Formula Weight 129.16
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material (1S,3R)-(+)-3-Aminocyclopentanecarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only