missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(+)-1-Amino-2-indanol 98.0+%, TCI America™

Catalog No. A23071G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23071G 1 g
A23075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A23071G Supplier TCI America Supplier No. A23071G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 163061-74-3
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD08275383
InChI Key LOPKSXMQWBYUOI-IUCAKERBSA-N
Synonym (1S,2S)-(+)-1-Amino-2-hydroxyindan
PubChem CID 6931156
IUPAC Name (1S,2S)-1-amino-2,3-dihydro-1H-inden-2-ol
SMILES C1C(C(C2=CC=CC=C21)N)O

Specifications

Melting Point 146°C
Color White-Yellow
Quantity 1 g
Formula Weight 149.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1S,2S)-(+)-1-Amino-2-indanol
EINECSNumber (4)-1717
TSCA No