missing translation for 'onlineSavingsMsg'
Learn More

(1S,2R)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine 98.0+%, TCI America™

Catalog No. B34831G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34831G 1 g
B34835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34831G Supplier TCI America Supplier No. B34831G
Only null left

Chemical Identifiers

CAS 365996-30-1
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
MDL Number MFCD09952105
InChI Key AKVIZYGPJIWKOS-BDAKNGLRSA-N
Synonym (1S,2R)-N1-Boc-1,2-cyclohexanediamine, (1S,2R)-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane, (1S,2R)-2-Amino-1-(Boc-amino)cyclohexane
PubChem CID 1514389
IUPAC Name tert-butyl N-[(1S,2R)-2-aminocyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCCCC1N

Specifications

Color White
Quantity 1 g
Formula Weight 214.31
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material (1S,2R)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine