accessibility menu, dialog, popup

UserName

H3CH3CCH3OONHH2N

CAS RN 365996-30-1

IUPAC Name: tert-butyl N-(2-aminocyclohexyl)carbamate
Synonyms: tert-butyl N-(2-aminocyclohexyl)carbamate
Molecular Weight (g/mol): 214.31
Molecular Formula: C11H22N2O2
InChi Key: AKVIZYGPJIWKOS-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC1CCCCC1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results