Learn More
(1S,2R)-(-)-cis-1-Amino-2-indanol, 99%
CAS: 126456-43-7 | C9H12NO | 150.20 g/mol
$45.33 - $45.33
Chemical Identifiers
| CAS | 126456-43-7 |
|---|---|
| Molecular Formula | C9H12NO |
| Molecular Weight (g/mol) | 150.20 |
| MDL Number | MFCD00216655 |
| InChI Key | LOPKSXMQWBYUOI-BDAKNGLRSA-O |
| PubChem CID | 9866743 |
| SMILES | [NH3+][C@@H]1[C@H](O)CC2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC315652500
|
Thermo Scientific Chemicals
315652500 |
250 mg | Glass bottle |
Each for $45.33
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 126456-43-7 | |
| 150.20 | |
| LOPKSXMQWBYUOI-BDAKNGLRSA-O | |
| [NH3+][C@@H]1[C@H](O)CC2=CC=CC=C12 |
| C9H12NO | |
| MFCD00216655 | |
| 9866743 |
Specifications
| 126456-43-7 | |
| 100.0 | |
| White to Beige | |
| 98.5% min. (GC) | |
| C9H12NO | |
| 250 mg | |
| 1s,2r-1-amino-2,3-dihydro-1h-inden-2-ol, 1s,2r---cis-1-amino-2-indanol, 1s,2r---1-amino-2-indanol, 1s,2r-1-amino-2-indanol, 1s,2r-cis-1-amino-2-indanol, cis-1s,2r-1-amino-2-indanol, 1s,2r-1-amino-indan-2-ol, 1s,2r---cis-1-aminoindan-2-ol, 1s,2r---1-amino-2-hydroxyindan, 1s, 2r-cis-1-amino-2-indanol | |
| LOPKSXMQWBYUOI-BDAKNGLRSA-O | |
| (1S,2R)-1-amino-2,3-dihydro-1H-inden-2-ol | |
| 9866743 | |
| 99% | |
| (1S, 2R)-(-)-cis-1-Amino-2-indanol, 99% |
| 98.5 | |
| 117°C to 121°C | |
| Authentic | |
| Glass bottle | |
| MFCD00216655 | |
| -62 | |
| Solubility in water: slightly soluble | |
| [NH3+][C@@H]1[C@H](O)CC2=CC=CC=C12 | |
| 150.20 | |
| 149.19 | |
| Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
TSCA : TSCA
RUO – Research Use Only