missing translation for 'onlineSavingsMsg'
Learn More

(1S,2R)-(-)-cis-1-Amino-2-indanol, 97%

Catalog No. p-7031109
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3206606 5 g
AAH3206614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3206606 Supplier Thermo Scientific Chemicals Supplier No. H3206606
Only null left

CAS: 126456-43-7 | C9H12NO | 150.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 126456-43-7
Molecular Formula C9H12NO
Molecular Weight (g/mol) 150.20
MDL Number MFCD00216655
InChI Key LOPKSXMQWBYUOI-BDAKNGLRSA-O
Synonym 1s,2r-1-amino-2,3-dihydro-1h-inden-2-ol, 1s,2r---cis-1-amino-2-indanol, 1s,2r---1-amino-2-indanol, 1s,2r-1-amino-2-indanol, 1s,2r-cis-1-amino-2-indanol, cis-1s,2r-1-amino-2-indanol, 1s,2r-1-amino-indan-2-ol, 1s,2r---cis-1-aminoindan-2-ol, 1s,2r---1-amino-2-hydroxyindan, 1s, 2r-cis-1-amino-2-indanol
PubChem CID 9866743
IUPAC Name (1S,2R)-1-amino-2,3-dihydro-1H-inden-2-ol
SMILES [NH3+][C@@H]1[C@H](O)CC2=CC=CC=C12

Specifications

Melting Point 117°C to 118°C
Quantity 5 g
UN Number UN3259
Sensitivity Air Sensitive
Optical Rotation −61° (c=0.5 in Chloroform)
Formula Weight 149.19
Percent Purity 97%
Chemical Name or Material (1S,2R)-(-)-cis-1-Amino-2-indanol
Hazard Category H302-H312-H315-H319-H332-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber 603-144-8
RTECSNumber NK7525500
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only