missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)propan-1-ol 98.0+%, TCI America™
Supplier: TCI America P13755G
Specifications
| (1S,2R)-1-Phenyl-2-(1-pyrrolidinyl)propan-1-ol | |
| 48°C | |
| C13H19NO | |
| 5 g | |
| FZVHJGJBJLFWEX-UHFFFAOYNA-N | |
| 1-phenyl-2-(pyrrolidin-1-yl)propan-1-ol | |
| 9942483 | |
| ≥98.0% (GC) |
| 123620-80-4 | |
| White-Yellow | |
| MFCD02093492 | |
| 1s,2r-1-phenyl-2-pyrrolidin-1-yl propan-1-ol, 1s,2r-1-phenyl-2-1-pyrrolidinyl propan-1-ol, 1s,2r-1-phenyl-2-pyrrolidin-1-yl-propan-1-ol, 1s,2r-1-phenyl-2-1-pyrrolidinyl-1-propanol, 1s,2r-n,n-tetramethylenenorephedrine, 1-phenyl-2-pyrrolidin-1-yl propan-1-ol, 1s,2r, 1-pyrrolidineethanol, beta-methyl-alpha-phenyl-, alphas,betar, pubchem8276, pubchem14465, 1s,2r-1-phenyl-2-pyrrolizino-1-propanol | |
| CC(C(O)C1=CC=CC=C1)N1CCCC1 | |
| 205.30 | |
| 205.30 | |
| Crystalline Powder |
Chemical Identifiers
| 123620-80-4 | |
| 205.30 | |
| FZVHJGJBJLFWEX-UHFFFAOYNA-N | |
| 9942483 | |
| CC(C(O)C1=CC=CC=C1)N1CCCC1 |
| C13H19NO | |
| MFCD02093492 | |
| 1s,2r-1-phenyl-2-pyrrolidin-1-yl propan-1-ol, 1s,2r-1-phenyl-2-1-pyrrolidinyl propan-1-ol, 1s,2r-1-phenyl-2-pyrrolidin-1-yl-propan-1-ol, 1s,2r-1-phenyl-2-1-pyrrolidinyl-1-propanol, 1s,2r-n,n-tetramethylenenorephedrine, 1-phenyl-2-pyrrolidin-1-yl propan-1-ol, 1s,2r, 1-pyrrolidineethanol, beta-methyl-alpha-phenyl-, alphas,betar, pubchem8276, pubchem14465, 1s,2r-1-phenyl-2-pyrrolizino-1-propanol | |
| 1-phenyl-2-(pyrrolidin-1-yl)propan-1-ol |
Safety and Handling
TSCA : No