missing translation for 'onlineSavingsMsg'
Learn More

(1S,2R)-(-)-1-Amino-2-indanol 98.0+%, TCI America™

Catalog No. A16241G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16241G 1 g
A16245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16241G Supplier TCI America Supplier No. A16241G

Chemical Identifiers

CAS 126456-43-7
Molecular Formula C9H12NO
Molecular Weight (g/mol) 150.20
MDL Number MFCD00216655
InChI Key LOPKSXMQWBYUOI-BDAKNGLRSA-O
Synonym 1s,2r-1-amino-2,3-dihydro-1h-inden-2-ol, 1s,2r---cis-1-amino-2-indanol, 1s,2r---1-amino-2-indanol, 1s,2r-1-amino-2-indanol, 1s,2r-cis-1-amino-2-indanol, cis-1s,2r-1-amino-2-indanol, 1s,2r-1-amino-indan-2-ol, 1s,2r---cis-1-aminoindan-2-ol, 1s,2r---1-amino-2-hydroxyindan, 1s, 2r-cis-1-amino-2-indanol
PubChem CID 9866743
IUPAC Name (1S,2R)-2-hydroxy-2,3-dihydro-1H-inden-1-aminium
SMILES [NH3+][C@@H]1[C@H](O)CC2=CC=CC=C12

Specifications

Melting Point 117°C
Quantity 1 g
Formula Weight 149.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (1S,2R)-(-)-1-Amino-2-indanol
EINECSNumber (4)-1717
TSCA No