missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S)-(-)-10-Mercaptoborneol 97.0+%, TCI America™
Supplier: TCI America M1341100MG
Specifications
| (1S)-(-)-10-Mercaptoborneol | |
| 86°C | |
| C10H18OS | |
| 100 mg | |
| CC1(C)C2CCC1(CS)C(O)C2 | |
| 186.31 | |
| 186.31 | |
| Crystalline Powder |
| 71242-59-6 | |
| White | |
| MFCD02093482 | |
| PYQMNINTTPIRIT-UHFFFAOYNA-N | |
| 7,7-dimethyl-1-(sulfanylmethyl)bicyclo[2.2.1]heptan-2-ol | |
| 130758660 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 71242-59-6 | |
| 186.31 | |
| PYQMNINTTPIRIT-UHFFFAOYNA-N | |
| 7,7-dimethyl-1-(sulfanylmethyl)bicyclo[2.2.1]heptan-2-ol |
| C10H18OS | |
| MFCD02093482 | |
| 130758660 | |
| CC1(C)C2CCC1(CS)C(O)C2 |
Safety and Handling
TSCA : No