CAS RN 71242-59-6
CAS RN 71242-59-6
IUPAC Name:
7,7-dimethyl-1-(sulfanylmethyl)bicyclo[2.2.1]heptan-2-ol
Synonyms:
7,7-dimethyl-1-(sulfanylmethyl)bicyclo[2.2.1]heptan-2-ol
Molecular Weight (g/mol):
186.31
Molecular Formula:
C10H18OS
InChi Key:
PYQMNINTTPIRIT-UHFFFAOYNA-N
SMILES:
CC1(C)C2CCC1(CS)C(O)C2
1
–
1
of
1
results
1
(1S)-(-)-10-Mercaptoborneol 97.0+%, TCI America™
CAS: 71242-59-6 Molecular Formula: C10H18OS Molecular Weight (g/mol): 186.31 MDL Number: MFCD02093482 InChI Key: PYQMNINTTPIRIT-UHFFFAOYNA-N PubChem CID: 130758660 IUPAC Name: 7,7-dimethyl-1-(sulfanylmethyl)bicyclo[2.2.1]heptan-2-ol SMILES: CC1(C)C2CCC1(CS)C(O)C2