missing translation for 'onlineSavingsMsg'
Learn More

(1RS)-1-(6-Methoxynaphthalen-2-yl)ethanol Pharmaceutical Impurity Standard, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015493
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015493 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015493 Supplier Merck Emd Millipore Supplier No. 8071350MG
Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 77301-42-9
Linear Formula C13H14O2
MDL Number MFCD01632581
Synonym Naproxen impurity K (PhEur); alpha-(6-Methoxy-2-naphthyl)ethanol; (+/-)-1-(6-Methoxy-2-naphthyl)ethanol; 1-[6-Methoxy-2-naphthyl]hydroxyethane; 2-(1-Hydroxyethyl)-6-methoxynaphthalene; 6-Methoxy-alpha-methyl-2-naphthalenemethanol
Type Pharmaceutical Impurity Standard
Quantity 50 mg
Formula Weight 202.25
Percent Purity ≥98.0% (HPLC)