missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R)-(-)-Thiocamphor 97.0+%, TCI America™
$180.97 - $550.61
Chemical Identifiers
| CAS | 53402-10-1 |
|---|---|
| Molecular Formula | C10H16S |
| Molecular Weight (g/mol) | 168.298 |
| MDL Number | MFCD00074855 |
| InChI Key | AAADKYXUTOBAGS-MHPPCMCBSA-N |
| PubChem CID | 12470051 |
| IUPAC Name | (4R)-4,7,7-trimethylbicyclo[2.2.1]heptane-3-thione |
| SMILES | CC1(C2CCC1(C(=S)C2)C)C |
Chemical Identifiers
| 53402-10-1 | |
| 168.298 | |
| AAADKYXUTOBAGS-MHPPCMCBSA-N | |
| (4R)-4,7,7-trimethylbicyclo[2.2.1]heptane-3-thione |
| C10H16S | |
| MFCD00074855 | |
| 12470051 | |
| CC1(C2CCC1(C(=S)C2)C)C |
Specifications
| 53402-10-1 | |
| C10H16S | |
| 1 g | |
| CC1(C2CCC1(C(=S)C2)C)C | |
| 168.298 | |
| 168.30 | |
| Crystalline Powder |
| 137°C | |
| MFCD00074855 | |
| AAADKYXUTOBAGS-MHPPCMCBSA-N | |
| (4R)-4,7,7-trimethylbicyclo[2.2.1]heptane-3-thione | |
| 12470051 | |
| ≥97.0% (GC) | |
| (1R)-(-)-Thiocamphor |
Safety and Handling
TSCA : No