accessibility menu, dialog, popup

UserName

H3CSCH3H3C

CAS RN 53402-10-1

IUPAC Name: 1,7,7-trimethylbicyclo[2.2.1]heptane-2-thione
Synonyms: thiocamphor
Molecular Weight (g/mol): 168.3
Molecular Formula: C10H16S
InChi Key: AAADKYXUTOBAGS-UHFFFAOYNA-N
SMILES: CC1(C)C2CCC1(C)C(=S)C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results