missing translation for 'onlineSavingsMsg'
Learn More

(1R)-(-)-Thiocamphor 97.0+%, TCI America™

Catalog No. T18631G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18631G 1 g
T18635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18631G Supplier TCI America Supplier No. T18631G
Only null left

Chemical Identifiers

CAS 53402-10-1
Molecular Formula C10H16S
Molecular Weight (g/mol) 168.298
MDL Number MFCD00074855
InChI Key AAADKYXUTOBAGS-MHPPCMCBSA-N
PubChem CID 12470051
IUPAC Name (4R)-4,7,7-trimethylbicyclo[2.2.1]heptane-3-thione
SMILES CC1(C2CCC1(C(=S)C2)C)C

Specifications

Melting Point 137°C
Quantity 1 g
Formula Weight 168.30
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1R)-(-)-Thiocamphor