Learn More
(1R)-(-)-Menthyl glyoxylate monohydrate, 98%
CAS: 111969-64-3 | C12H22O4 | 230.304 g/mol
$155.52 - $441.33
Chemical Identifiers
| CAS | 111969-64-3 |
|---|---|
| Molecular Formula | C12H22O4 |
| Molecular Weight (g/mol) | 230.304 |
| MDL Number | MFCD00792503 |
| InChI Key | BWZMJRSMHQDFIT-KXUCPTDWSA-N |
| Synonym | 1r,2s,5r-2-isopropyl-5-methylcyclohexyl 2,2-dihydroxyacetate, l-menthyl glyoxylate hydrate, 1r,2s,5r-5-methyl-2-1-methylethyl cyclohexyl dihydroxy-acetate, l-menthyl glyoxylate, l-menthylglyoxylate hydrate, --menthyl 2,2-dihydroxyacetate, 1r,2s,5r-5-methyl-2-1-methylethyl cyclohexyl di, 2,2-dihydroxyacetic acid 1r,3r,4s-p-menth-3-yl ester, 1r,2s,5r-5-methyl-2-propan-2-ylcyclohexyl 2,2-dihydroxyacetate, 2,2-dihydroxyacetic acid 1r,2s,5r-2-isopropyl-5-methylcyclohexyl ester |
| PubChem CID | 7373179 |
| IUPAC Name | [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] 2,2-dihydroxyacetate |
| SMILES | CC1CCC(C(C1)OC(=O)C(O)O)C(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1920814
|
Thermo Scientific Chemicals
L1920814 |
25 g |
N/A
|
|
|||||
|
AAL1920822
|
Thermo Scientific Chemicals
L1920822 |
100 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 111969-64-3 | |
| 230.304 | |
| BWZMJRSMHQDFIT-KXUCPTDWSA-N | |
| 7373179 | |
| CC1CCC(C(C1)OC(=O)C(O)O)C(C)C |
| C12H22O4 | |
| MFCD00792503 | |
| 1r,2s,5r-2-isopropyl-5-methylcyclohexyl 2,2-dihydroxyacetate, l-menthyl glyoxylate hydrate, 1r,2s,5r-5-methyl-2-1-methylethyl cyclohexyl dihydroxy-acetate, l-menthyl glyoxylate, l-menthylglyoxylate hydrate, --menthyl 2,2-dihydroxyacetate, 1r,2s,5r-5-methyl-2-1-methylethyl cyclohexyl di, 2,2-dihydroxyacetic acid 1r,3r,4s-p-menth-3-yl ester, 1r,2s,5r-5-methyl-2-propan-2-ylcyclohexyl 2,2-dihydroxyacetate, 2,2-dihydroxyacetic acid 1r,2s,5r-2-isopropyl-5-methylcyclohexyl ester | |
| [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] 2,2-dihydroxyacetate |
Specifications
| 111969-64-3 | |
| 1.182 | |
| C12H22O4 | |
| 25 g | |
| 5637371 | |
| 1r,2s,5r-2-isopropyl-5-methylcyclohexyl 2,2-dihydroxyacetate, l-menthyl glyoxylate hydrate, 1r,2s,5r-5-methyl-2-1-methylethyl cyclohexyl dihydroxy-acetate, l-menthyl glyoxylate, l-menthylglyoxylate hydrate, --menthyl 2,2-dihydroxyacetate, 1r,2s,5r-5-methyl-2-1-methylethyl cyclohexyl di, 2,2-dihydroxyacetic acid 1r,3r,4s-p-menth-3-yl ester, 1r,2s,5r-5-methyl-2-propan-2-ylcyclohexyl 2,2-dihydroxyacetate, 2,2-dihydroxyacetic acid 1r,2s,5r-2-isopropyl-5-methylcyclohexyl ester | |
| −75° (c=1 in Methanol) | |
| [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] 2,2-dihydroxyacetate | |
| 7373179 | |
| 98% |
| 76°C to 80°C | |
| 95°C to 96°C (0.3 mmHg) | |
| MFCD00792503 | |
| UN3077 | |
| Air Sensitive | |
| BWZMJRSMHQDFIT-KXUCPTDWSA-N | |
| CC1CCC(C(C1)OC(=O)C(O)O)C(C)C | |
| 230.304 | |
| 230.31 (212.29 Anhydrous) | |
| (1R)-(-)-Menthyl glyoxylate monohydrate |
Safety and Handling
GHS H Statement
H318-H373
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
P260-P280-P305+P351+P338-P310-P314-P501c
H318-H373
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 416-810-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only