accessibility menu, dialog, popup

UserName

CAS RN 111969-64-3

IUPAC Name: 5-methyl-2-(propan-2-yl)cyclohexyl 2,2-dihydroxyacetate
Synonyms: 2-isopropyl-5-methylcyclohexyl 2,2-dihydroxyacetate
Molecular Weight (g/mol): 230.3
Molecular Formula: C12H22O4
InChi Key: BWZMJRSMHQDFIT-UHFFFAOYNA-N
SMILES: CC(C)C1CCC(C)CC1OC(=O)C(O)O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)