missing translation for 'onlineSavingsMsg'
Learn More

(1R)-(-)-Menthyl glyoxylate monohydrate, 98%

Catalog No. AAL1920814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1920814 25 g
AAL1920822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1920814 Supplier Thermo Scientific Chemicals Supplier No. L1920814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 111969-64-3 | C12H22O4 | 230.304 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111969-64-3
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.304
MDL Number MFCD00792503
InChI Key BWZMJRSMHQDFIT-KXUCPTDWSA-N
Synonym 1r,2s,5r-2-isopropyl-5-methylcyclohexyl 2,2-dihydroxyacetate, l-menthyl glyoxylate hydrate, 1r,2s,5r-5-methyl-2-1-methylethyl cyclohexyl dihydroxy-acetate, l-menthyl glyoxylate, l-menthylglyoxylate hydrate, --menthyl 2,2-dihydroxyacetate, 1r,2s,5r-5-methyl-2-1-methylethyl cyclohexyl di, 2,2-dihydroxyacetic acid 1r,3r,4s-p-menth-3-yl ester, 1r,2s,5r-5-methyl-2-propan-2-ylcyclohexyl 2,2-dihydroxyacetate, 2,2-dihydroxyacetic acid 1r,2s,5r-2-isopropyl-5-methylcyclohexyl ester
PubChem CID 7373179
IUPAC Name [(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl] 2,2-dihydroxyacetate
SMILES CC1CCC(C(C1)OC(=O)C(O)O)C(C)C

Specifications

Melting Point 76°C to 80°C
Density 1.182
Boiling Point 95°C to 96°C (0.3 mmHg)
Quantity 25 g
UN Number UN3077
Beilstein 5637371
Sensitivity Air Sensitive
Optical Rotation −75° (c=1 in Methanol)
Formula Weight 230.31 (212.29 Anhydrous)
Percent Purity 98%
Chemical Name or Material (1R)-(-)-Menthyl glyoxylate monohydrate
Show More Show Less
Hazard Category H318-H373
Hazard Statement GHS H Statement
H318-H373
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P280-P305+P351+P338-P310-P314-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 416-810-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only